APO456793495 Apollo Scientific Ltd

Methyl 4-hydroxy-3-methoxy-5-nitrobenzoate

  • MDL number MFCD06203667
  • Cas Number : 42590-00-1
  • Molecular Weight : 227.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 159-161
Iupac Name methyl 4-hydroxy-3-methoxy-5-nitrobenzoate
Inchi Code 1S/C9H9NO6/c1-15-7-4-5(9(12)16-2)3-6(8(7)11)10(13)14/h3-4,11H,1-2H3
Inchi Key ZQIHGTCQYHQBMY-UHFFFAOYSA-N

Description

Synonym 2-Hydroxy-3-methoxy-5-(methoxycarbonyl)nitrobenzene, 2-Methoxy-4-(methoxycarbonyl)-6-nitrophenol, 2-Hydroxy-5-(methoxycarbonyl)-3-nitroanisole, Methyl 4-hydroxy-5-nitro-m-anisate
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 159-161
Product Link https://store.apolloscientific.co.uk/product/methyl-4-hydroxy-3-methoxy-5-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-hydroxy-3-methoxy-5-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart