APO456793555 Apollo Scientific Ltd

3,6-Dichloropyridine-2-carboxamide

  • MDL number MFCD00186645
  • Cas Number : 1532-25-8
  • Molecular Weight : 191.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 178-180
Iupac Name 3,6-dichloro-2-pyridinecarboxamide
Inchi Code 1S/C6H4Cl2N2O/c7-3-1-2-4(8)10-5(3)6(9)11/h1-2H,(H2,9,11)
Inchi Key RFBSMVHKAFPDKD-UHFFFAOYSA-N

Description

Synonym 2-Carbamoyl-3,6-dichloropyridine, 3,6-Dichloropicolinamide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 178-180
Product Link https://store.apolloscientific.co.uk/product/36-dichloropyridine-2-carboxamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/36-dichloropyridine-2-carboxamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart