APO456793585 Apollo Scientific Ltd

4-Iodopiperidine, N-BOC protected

  • MDL number MFCD04115041
  • Cas Number : 301673-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-49
Synonym 1-(tert-Butoxycarbonyl)-4-iodopiperidine, tert-Butyl 4-iodopiperidine-1-carboxylate
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48653

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48653
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart