APO456793652 Apollo Scientific Ltd

5-Amino-3-bromo-1-methyl-1H-indazole

  • MDL number MFCD11100026
  • Cas Number : 1092351-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-1-methyl-1H-indazol-5-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-bromo-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-bromo-1-methyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart