APO456793687 Apollo Scientific Ltd

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzyl alcohol

  • MDL number MFCD07368531
  • Cas Number : 852180-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [4-(5-Methyl-1,2,4-oxadiazol-3-yl)phenyl]methanol, 3-[4-(Hydroxymethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-5-methyl-124-oxadiazol-3-ylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/4-5-methyl-124-oxadiazol-3-ylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart