APO456793766 Apollo Scientific Ltd

1,5-Anhydro-D-glucitol

  • MDL number MFCD00067386
  • Cas Number : 154-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-143
Synonym 1,5-Anhydro-D-sorbitol, (2R,3S,4R,5S)-2-(Hydroxymethyl)tetrahydropyran-3,4,5-triol
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12850

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12850
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart