APO456793784 Apollo Scientific Ltd

2-(Chloromethyl)-4-hydroxyquinazoline

  • MDL number MFCD00665825
  • Cas Number : 3817-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249-253
Synonym 2-(Chloromethyl)quinazlin-4-ol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloromethyl-4-hydroxyquinazoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloromethyl-4-hydroxyquinazoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart