APO456793786 Apollo Scientific Ltd

2,1,3-Benzoxadiazole-4-sulphonyl chloride

  • MDL number MFCD00068072
  • Cas Number : 114322-14-4
  • Molecular Weight : 218.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 77-78
Iupac Name 2,1,3-benzoxadiazole-4-sulfonyl chloride
Inchi Code 1S/C6H3ClN2O3S/c7-13(10,11)5-3-1-2-4-6(5)9-12-8-4/h1-3H
Inchi Key SIYWVPMYOGMRDW-UHFFFAOYSA-N

Description

Synonym Benzofurazan-4-sulphonyl chloride, 4-(Chlorosulphonyl)-2,1,3-benzoxadiazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 77-78
Product Link https://store.apolloscientific.co.uk/product/213-benzoxadiazole-4-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/213-benzoxadiazole-4-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart