APO456793832 Apollo Scientific Ltd

Disodium 7-aminonaphthalene-1,3-disulphonate

  • MDL number MFCD07773075
  • Cas Number : 842-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Aminonaphthalene-1,3-disulphonic acid, disodium salt
Purity 80
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30609

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30609
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart