APO456793845 Apollo Scientific Ltd

2-(Aminomethyl)-5-hydroxypyridine hydrochloride

  • MDL number MFCD17214179
  • Cas Number : 1276056-66-6
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-(Aminomethyl)pyridin-3-ol hydrochloride, (5-Hydroxypyridin-2-yl)methylamine hydrochloride
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-5-hydroxypyridine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-5-hydroxypyridine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart