APO456793850 Apollo Scientific Ltd

3-Hydroxy-4-methoxypyridine

  • MDL number MFCD11044354
  • Cas Number : 153199-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Synonym 4-Methoxypyridin-3-ol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-4-methoxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-4-methoxypyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart