APO456794154 Apollo Scientific Ltd

4-Hydroxyphthalic acid

  • MDL number MFCD00013984
  • Cas Number : 610-35-5
  • Molecular Weight : 182.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 196-198
Iupac Name 4-hydroxyphthalic acid
Inchi Code 1S/C8H6O5/c9-4-1-2-5(7(10)11)6(3-4)8(12)13/h1-3,9H,(H,10,11)(H,12,13)
Inchi Key MWRVRCAFWBBXTL-UHFFFAOYSA-N

Description

Synonym 3,4-Dicarboxyphenol, 4-Hydroxy-1,2-benzenedicarboxylic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 196-198
Product Link https://store.apolloscientific.co.uk/product/4-hydroxyphthalic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxyphthalic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart