APO456794188 Apollo Scientific Ltd

Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate

  • MDL number MFCD08704625
  • Cas Number : 425392-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56
Synonym 2-Bromo-5-chloro-4-(ethoxycarbonyl)-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-bromo-5-chloro-13-thiazole-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-bromo-5-chloro-13-thiazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart