APO456794191 Apollo Scientific Ltd

2-Bromo-3-(bromomethyl)pyridine

  • MDL number MFCD08276299
  • Cas Number : 94446-97-6
  • Molecular Weight : 250.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromonicotinyl bromide, alpha,2-Dibromo-3-picoline
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-bromomethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-bromomethylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart