APO456794220 Apollo Scientific Ltd

Ethyl 2-sulphanyl-1H-imidazole-4-carboxylate

  • MDL number MFCD00068042
  • Cas Number : 64038-64-8
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 191-193
Iupac Name ethyl 2-sulfanyl-1H-imidazole-4-carboxylate
Inchi Code 1S/C6H8N2O2S/c1-2-10-5(9)4-3-7-6(11)8-4/h3H,2H2,1H3,(H2,7,8,11)
Inchi Key PONOGPICUOALRU-UHFFFAOYSA-N

Description

Synonym Ethyl 2-mercapto-1H-imidazole-4-carboxylate, 4-(Ethoxycarbonyl)-2-sulphanyl-1H-imidazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 191-193
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-sulphanyl-1h-imidazole-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-sulphanyl-1h-imidazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart