APO456794238 Apollo Scientific Ltd

8-Bromo-6-methylimidazo[1,2-a]pyridine

  • MDL number MFCD09258770
  • Cas Number : 136117-93-6
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 62-64
Iupac Name 8-bromo-6-methylimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7BrN2/c1-6-4-7(9)8-10-2-3-11(8)5-6/h2-5H,1H3
Inchi Key IEXMXPBKRWDOBC-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-64
Product Link https://store.apolloscientific.co.uk/product/8-bromo-6-methylimidazo12-apyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-6-methylimidazo12-apyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart