APO456794351 Apollo Scientific Ltd

4-(tert-Butyl)phenylacetonitrile

  • MDL number MFCD00128112
  • Cas Number : 3288-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [4-(tert-Butyl)phenyl]acetonitrile, 4-(tert-Butyl)benzyl cyanide
Purity 98
Density 0.962
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 141-142/13mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13568

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13568
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart