APO456794385 Apollo Scientific Ltd

2-Bromo-5-chlorobenzonitrile

  • MDL number MFCD08445660
  • Cas Number : 57381-37-0
  • Molecular Weight : 216.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 138-139
Iupac Name 2-bromo-5-chlorobenzonitrile
Inchi Code 1S/C7H3BrClN/c8-7-2-1-6(9)3-5(7)4-10/h1-3H
Inchi Key CTSHRMBLMKPDAG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 138-139
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-chlorobenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-chlorobenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart