APO456794387 Apollo Scientific Ltd

Thieno[2,3-b]pyridine-2-carboxaldehyde

  • MDL number MFCD07772791
  • Cas Number : 53174-98-4
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 141.5-143
Iupac Name thieno[2,3-b]pyridine-2-carbaldehyde
Inchi Code 1S/C8H5NOS/c10-5-7-4-6-2-1-3-9-8(6)11-7/h1-5H
Inchi Key HGZQKYOLRLVMHZ-UHFFFAOYSA-N

Description

Synonym 2-Formylthieno[2,3-b]pyridine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 141.5-143
Product Link https://store.apolloscientific.co.uk/product/thieno23-bpyridine-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/thieno23-bpyridine-2-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart