APO456794445 Apollo Scientific Ltd

3-Bromo-5-methylbenzene-1,2-diamine

  • MDL number MFCD18088524
  • Cas Number : 70733-25-4
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 70-72
Iupac Name 3-bromo-5-methyl-1,2-benzenediamine
Inchi Code 1S/C7H9BrN2/c1-4-2-5(8)7(10)6(9)3-4/h2-3H,9-10H2,1H3
Inchi Key WHVCTNJQZXBGDH-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-methylphenylene-1,2-diamine, 3-Bromo-4,5-diaminotoluene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70-72
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methylbenzene-12-diamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methylbenzene-12-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart