APO456794581 Apollo Scientific Ltd

2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate

  • MDL number MFCD08166750
  • Cas Number : 1171826-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112(dec.)
Purity 95
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methylsulphonylbenzaldehyde-hydrate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methylsulphonylbenzaldehyde-hydrate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart