APO456794698 Apollo Scientific Ltd

1H-Indole-2-carbohydrazide

  • MDL number MFCD02293210
  • Cas Number : 5055-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243-245
Synonym 1H-Indole-2-carboxylic acid hydrazide, 2-(Hydrazinocarbonyl)-1H-indole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36676

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36676
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart