APO456794712 Apollo Scientific Ltd

1,4-Benzodioxan-2-carbonyl chloride

  • MDL number MFCD02180342
  • Cas Number : 3663-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-59
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12923

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12923
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart