APO456794721 Apollo Scientific Ltd

4-(Aminomethyl)-1-methyl-1H-indole

  • MDL number MFCD08690264
  • Cas Number : 864264-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-47
Synonym 1-(1-Methyl-1H-indol-4-yl)methylamine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32689

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32689
pictograms GHS05;GHS07
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart