APO456794770 Apollo Scientific Ltd

1-Bromo-8-phenyloctane

  • MDL number MFCD01075178
  • Cas Number : 54646-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (8-Bromooct-1-yl)benzene
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 84-86/0.15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13406

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13406
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart