APO456794951 Apollo Scientific Ltd

4-Bromothiophene-3-boronic acid

  • MDL number MFCD06801663
  • Cas Number : 101084-76-8
  • Molecular Weight : 206.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-198
Synonym 3-Bromothiophene-4-boronic acid
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromothiophene-3-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromothiophene-3-boronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart