APO456795016 Apollo Scientific Ltd

3-Methyl-4-nitro-1H-pyrazole

  • MDL number MFCD00037864
  • Cas Number : 5334-39-4
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-134
Synonym 5-Methyl-4-nitro-1H-pyrazole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 325
Product Link https://store.apolloscientific.co.uk/product/3-methyl-4-nitro-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-4-nitro-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart