APO456795035 Apollo Scientific Ltd

2'-Bromo-5'-methoxyacetophenone

  • MDL number MFCD02683853
  • Cas Number : 6342-63-8
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Bromo-5-methoxyphenyl)ethan-1-one, 3-Acetyl-4-bromoanisole
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-methoxyacetophenone

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-methoxyacetophenone
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart