APO456795063 Apollo Scientific Ltd

5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD00275475
  • Cas Number : 18735-74-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-183.5
Synonym 4-Carboxy-5-methyl-2-phenyl-1,3-oxazole, (4-Carboxy-5-methyl-1,3-oxazol-2-yl)benzene
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-2-phenyl-13-oxazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-2-phenyl-13-oxazole-4-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart