APO456795070 Apollo Scientific Ltd

1-Methyl-3-phenyl-1H-pyrazole-5-carboxaldehyde

  • MDL number MFCD08271941
  • Cas Number : 864068-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Synonym 5-Formyl-1-methyl-3-phenyl-1H-pyrazole
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32049

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32049
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart