APO456795090 Apollo Scientific Ltd

1-Methyl-1H-indole-4-carboxylic acid

  • MDL number MFCD08435874
  • Cas Number : 90924-06-4
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 243-245
Iupac Name 1-methyl-1H-indole-4-carboxylic acid
Inchi Code 1S/C10H9NO2/c1-11-6-5-7-8(10(12)13)3-2-4-9(7)11/h2-6H,1H3,(H,12,13)
Inchi Key FUPJJUHCLDXVGX-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-1-methyl-1H-indole
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 243-245
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P260;P271
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart