APO456795112 Apollo Scientific Ltd

3-(Thien-2-yl)-1H-pyrazole

  • MDL number MFCD00178757
  • Cas Number : 19933-24-5
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-thien-2-yl-1h-pyrazole

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/3-thien-2-yl-1h-pyrazole
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart