APO456795179 Apollo Scientific Ltd

6-Methylpyridine-2-boronic acid

  • MDL number MFCD03093331
  • Cas Number : 372963-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Ship on Ice
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32987

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=32987
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart