APO456795287 Apollo Scientific Ltd

5,7-Difluoro-2-oxindole

  • MDL number MFCD01075219
  • Cas Number : 247564-59-6
  • Molecular Weight : 169.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 220-222
Iupac Name 5,7-difluoro-1,3-dihydro-2H-indol-2-one
Inchi Code 1S/C8H5F2NO/c9-5-1-4-2-7(12)11-8(4)6(10)3-5/h1-3,11-12H
Inchi Key KTBCCPUGFJNHOP-UHFFFAOYSA-N

Description

Synonym 5,7-Difluoro-1,3-dihydro-2H-indol-2-one, 5,7-Difluoroindolin-2-one
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 220-222
Product Link https://store.apolloscientific.co.uk/product/57-difluoro-2-oxindole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/57-difluoro-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart