APO456795425 Apollo Scientific Ltd

3-Fluoro-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD01631565
  • Cas Number : 261951-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-121
Synonym alpha,alpha,alpha,3-Tetrafluoro-o-toluic acid, 2-Carboxy-6-fluorobenzotrifluoride
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart