APO456795459 Apollo Scientific Ltd

4-(Trifluoromethoxy)-DL-phenylglycine

  • MDL number MFCD01631461
  • Cas Number : 261952-24-3
  • Molecular Weight : 235.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name amino[4-(trifluoromethoxy)phenyl]acetic acid
Inchi Code 1S/C9H8F3NO3/c10-9(11,12)16-6-3-1-5(2-4-6)7(13)8(14)15/h1-4,7H,13H2,(H,14,15)
Inchi Key KBWQXEWTAXZMRX-UHFFFAOYSA-N

Description

Synonym Amino[4-(trifluoromethoxy)phenyl]acetic acid
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethoxy-dl-phenylglycine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethoxy-dl-phenylglycine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart