APO456795473 Apollo Scientific Ltd

Hexakis(1H,1H-perfluoropropoxy)phosphazene

  • MDL number MFCD00798141
  • Cas Number : 429-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 14-16
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 120-122/2.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3674

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3674
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart