APO456795567 Apollo Scientific Ltd

4-Fluorophenyl acetate

  • MDL number MFCD00041361
  • Cas Number : 405-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Acetoxy-4-fluorobenzene
Purity 98
Density 1.178
Flash Point 87
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 197-199
Product Link https://store.apolloscientific.co.uk/product/4-fluorophenyl-acetate

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluorophenyl-acetate
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart