APO456795591 Apollo Scientific Ltd

Ethyl 1H-tetrafluoropropanaote

  • MDL number MFCD04039197
  • Cas Number : 399-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 2,3,3,3-tetrafluoropropanoate, Ethyl 2,3,3,3-tetrafluoropropionate
Purity 98
Density 1.2884
Flash Point 18
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 108-109
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8577

Safety Information

signalword Danger
pcodes P280;P210
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8577
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart