APO456795641 Apollo Scientific Ltd

6-Bromo-3-fluoro-2-methylpyridine

  • MDL number MFCD03095064
  • Cas Number : 374633-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Synonym 6-Bromo-3-fluoro-2-picoline
Purity 98
Flash Point 65
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-fluoro-2-methylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-fluoro-2-methylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart