APO456795787 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethoxy)phenol

  • MDL number MFCD06660342
  • Cas Number : 177596-38-2
  • Molecular Weight : 196.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethoxyphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethoxyphenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart