APO456795837 Apollo Scientific Ltd

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid

  • MDL number MFCD00277482
  • Cas Number : 80194-68-9
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-138
Synonym 2-Carboxy-3-chloro-5-(trifluoromethyl)pyridine, 3-Chloro-5-(trifluoromethyl)picolinic acid
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-trifluoromethylpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart