APO456795955 Apollo Scientific Ltd

2,2-Difluoropropan-1-ol

  • MDL number MFCD07782150
  • Cas Number : 33420-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Difluoro-1-hydroxypropane
Density 1.1642
Flash Point 31.1
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 97-99
Product Link https://store.apolloscientific.co.uk/product/22-difluoropropan-1-ol

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluoropropan-1-ol
pictograms GHS02;GHS07
hstatements H226;H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart