APO456795965 Apollo Scientific Ltd

3-Chloro-2-iodo-1,1,1-trifluoropropane

  • MDL number MFCD01862016
  • Cas Number : 113402-77-0
  • Molecular Weight : 258.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-iodo-111-trifluoropropane

Safety Information

signalword Warning
pcodes P280;P302+P352;P261
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-iodo-111-trifluoropropane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart