APO456795966 Apollo Scientific Ltd

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine

  • MDL number MFCD00153072
  • Cas Number : 75806-84-7
  • Molecular Weight : 260.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-5-chloro-alpha,alpha,alpha-trifluoro-3-picoline
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80/32mm
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-5-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart