APO456796023 Apollo Scientific Ltd

4-(Difluoromethyl)benzeneboronic acid

  • MDL number MFCD09056705
  • Cas Number : 946525-43-5
  • Molecular Weight : 171.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Boronobenzal fluoride
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart