APO456796026 Apollo Scientific Ltd

1-Chloro-2,6-dibromo-4-fluorobenzene

  • MDL number MFCD00061122
  • Cas Number : 179897-90-6
  • Molecular Weight : 288.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 84-87
Iupac Name 1,3-dibromo-2-chloro-5-fluorobenzene
Inchi Code 1S/C6H2Br2ClF/c7-4-1-3(10)2-5(8)6(4)9/h1-2H
Inchi Key PZKDJJMHRYNBOR-UHFFFAOYSA-N

Description

Synonym 1,3-Dibromo-2-chloro-5-fluorobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 84-87
Product Link https://store.apolloscientific.co.uk/product/1-chloro-26-dibromo-4-fluorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-chloro-26-dibromo-4-fluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart