APO456796060 Apollo Scientific Ltd

3-Fluoro-5-isobutoxybenzeneboronic acid

  • MDL number MFCD07363783
  • Cas Number : 850589-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-134
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-isobutoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-isobutoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart