APO456796084 Apollo Scientific Ltd

N-Chloroacetyl-2-(trifluoromethyl)aniline

  • MDL number MFCD00018893
  • Cas Number : 3792-04-9
  • Molecular Weight : 237.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 2-Chloro-2'-(trifluoromethyl)acetanilide
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-chloroacetyl-2-trifluoromethylaniline

Safety Information

signalword Warning
pcodes P280;P321
MSDS https://store.apolloscientific.co.uk/product/n-chloroacetyl-2-trifluoromethylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart