APO456796103 Apollo Scientific Ltd

7-Fluoro-2-oxindole

  • MDL number MFCD02179608
  • Cas Number : 71294-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Synonym 1,3-Dihydro-7-fluoro-2H-indol-2-one, 7-Fluoro-2-indolone
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/7-fluoro-2-oxindole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-fluoro-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart